accessibility menu, dialog, popup

UserName

CAS RN 321674-73-1

H3CONHOOH(E/Z)

CAS RN 321674-73-1

IUPAC Name: 2-[3-(naphthalen-2-yl)but-2-enamido]benzoic acid
Synonyms: 2-[3-(naphthalen-2-yl)but-2-enamido]benzoic acid
Molecular Weight (g/mol): 331.37
Molecular Formula: C21H17NO3
InChi Key: PGFQXGLPJUCTOI-UHFFFAOYSA-N
SMILES: CC(=CC(=O)NC1=CC=CC=C1C(O)=O)C1=CC=C2C=CC=CC2=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results