accessibility menu, dialog, popup

UserName

H3COOBr

CAS RN 33626-98-1

IUPAC Name: methyl 6-bromonaphthalene-2-carboxylate
Synonyms: methyl 6-bromonaphthalene-2-carboxylate
Molecular Weight (g/mol): 265.11
Molecular Formula: C12H9BrO2
InChi Key: JEUBRLPXJZOGPX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2C=C(Br)C=CC2=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)