CAS RN 3374-88-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ICA, MedChemExpress
MedChemExpress ICA (N-(pyridin-2-yl)-4-(pyridin-2-yl)thiazol-2-amine) is a SK channel inhibitor that has antileishmanial activity with an IC50 of 2.1 μM.