CAS RN 348-51-6
IUPAC Name:
1-chloro-2-fluorobenzene
Synonyms:
1-chloro-2-fluorobenzene M-chlorofluorobenzene
Molecular Weight (g/mol):
130.55
Molecular Formula:
C6H4ClF
InChi Key:
ZCJAYDKWZAWMPR-UHFFFAOYSA-N
SMILES:
FC1=CC=CC=C1Cl
1
–
6
of
6
results
1
1-Chloro-2-fluorobenzene, 98+%
CAS: 348-51-6 Molecular Formula: C6H4ClF Molecular Weight (g/mol): 130.55 MDL Number: MFCD00000533 InChI Key: ZCJAYDKWZAWMPR-UHFFFAOYSA-N Synonym: 2-chlorofluorobenzene,o-chlorofluorobenzene,benzene, 1-chloro-2-fluoro,o-fluorochlorobenzene,1-fluoro-2-chlorobenzene,1-chloro-2-fluoro-benzene,benzene, chlorofluoro,2-chloro-1-fluorobenzene,chlorofluorobenzene,2-fluorochlorobenzene PubChem CID: 9583 IUPAC Name: 1-chloro-2-fluorobenzene SMILES: FC1=CC=CC=C1Cl