accessibility menu, dialog, popup

UserName

H3COOF

CAS RN 34837-84-8

IUPAC Name: methyl 2-(4-fluorophenyl)acetate
Synonyms: methyl 2-(4-fluorophenyl)acetate
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: AJPPKGMEHMXPMC-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)