missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluorophenylacetate, 98%

Product Code. 15494927
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494927 5 g 5g
15404937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494927 Supplier Thermo Scientific Alfa Aesar Supplier No. H32236.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34837-84-8 | C9H9FO2 | 168.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34837-84-8
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00800608
InChI Key AJPPKGMEHMXPMC-UHFFFAOYSA-N
Synonym methyl 2-4-fluorophenyl acetate, methyl 4-fluorophenylacetate, 4-fluorophenylacetic acid methyl ester, pubchem24389, acmc-209iaq, methyl-4-fluorphenylacetate, methyl-4-fluorophenylacetate, methyl 4-fluorophenyl-acetate, methyl 4-fluorophenyl acetate
PubChem CID 2733233
IUPAC Name methyl 2-(4-fluorophenyl)acetate
SMILES COC(=O)CC1=CC=C(C=C1)F

Specifications

Quantity 5 g
Formula Weight 168.17
Percent Purity 98%
Chemical Name or Material Methyl 4-fluorophenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.