missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluorophenylacetate, 99%, Thermo Scientific Chemicals

Product Code. 11956951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11956951 100 g Glass Bottle 100g
10781341 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11956951 Supplier Thermo Scientific Acros Supplier No. 304671000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34837-84-8 | Molecular Formula: C9H9FO2 | Molecular Weight: 168.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34837-84-8
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.06
InChI Key AJPPKGMEHMXPMC-UHFFFAOYSA-N
Synonym methyl 2-4-fluorophenyl acetate, methyl 4-fluorophenylacetate, 4-fluorophenylacetic acid methyl ester, pubchem24389, acmc-209iaq, methyl-4-fluorphenylacetate, methyl-4-fluorophenylacetate, methyl 4-fluorophenyl-acetate, methyl 4-fluorophenyl acetate
PubChem CID 2733233
IUPAC Name methyl 2-(4-fluorophenyl)acetate
SMILES COC(=O)CC1=CC=C(C=C1)F

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula FC6H4CH2CO2CH3
Refractive Index 1.4860 to 1.4880
Quantity 100 g
Formula Weight 168.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl 4-fluorophenylacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.