accessibility menu, dialog, popup

UserName

CAS RN 36861-47-9

CH3CH3OH3CH3C(E)

CAS RN 36861-47-9

IUPAC Name: (3E)-1,7,7-trimethyl-3-[(4-methylphenyl)methylidene]bicyclo[2.2.1]heptan-2-one
Synonyms: 4-methylbenzylidene camphor
Molecular Weight (g/mol): 254.37
Molecular Formula: C18H22O
InChi Key: HEOCBCNFKCOKBX-SDNWHVSQNA-N
SMILES: CC1=CC=C(\C=C2/C3CCC(C)(C2=O)C3(C)C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results