accessibility menu, dialog, popup

UserName

NH2NONO(R)HO(S)HO(S)OHNH(R)N

CAS RN 38819-10-2

IUPAC Name: 2-amino-9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Synonyms: 2-amino-9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Molecular Weight (g/mol): 283.24
Molecular Formula: C10H13N5O5
InChi Key: NYHBQMYGNKIUIF-FJFJXFQQSA-N
SMILES: NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results