missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosylguanine, Thermo Scientific™

Product Code. 17342491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17342491 500 mg 500mg
17332491 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17342491 Supplier Thermo Scientific Acros Supplier No. 468635000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 38819-10-2
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00065486
InChI Key NYHBQMYGNKIUIF-FJFJXFQQSA-N
Synonym Araguanosine; Ara-G
IUPAC Name 2-amino-9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
SMILES NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Crystalline Powder
Color White
Melting Point 225°C
Quantity 500 mg
Formula Weight 283.24
Chemical Name or Material 9-beta-D-Arabinofuranosylguanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.