CAS RN 39627-98-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-(2,6-Dimethylphenyl)pyridine-2-carboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2',6'-Picolinoxylidide, TRC
CAS: 39627-98-0 Molecular Formula: C14 H14 N2 O Molecular Weight (g/mol): 226.27 Synonym: N-(2,6-Dimethylphenyl)pyridine-2-carboxamide,Bupivacaine Hydrochloride Imp. A (EP),Mepivacaine Hydrochloride Imp. C (EP) IUPAC Name: N-(2,6-dimethylphenyl)pyridine-2-carboxamide SMILES: Cc1cccc(C)c1NC(=O)c2ccccn2