accessibility menu, dialog, popup

UserName

CAS RN 4179-19-5

CH3OCH3OCH3

CAS RN 4179-19-5

IUPAC Name: 1,3-dimethoxy-5-methylbenzene
Synonyms: 3,5-dimethoxytoluene
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: RIZBLVRXRWHLFA-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)