accessibility menu, dialog, popup

UserName

HONHOCl

CAS RN 41859-57-8

IUPAC Name: 4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
Synonyms: 4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
Molecular Weight (g/mol): 275.73
Molecular Formula: C15H14ClNO2
InChi Key: ZTLWJYCDAXUIBK-UHFFFAOYSA-N
SMILES: OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results