CAS RN 442908-10-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Vipadenant, MedChemExpress
MedChemExpress Vipadenant (BIIB-014; CEB-4520) is an adenosine receptor antagonist, with Kis of 1.3 nM and 68 nM for A2A and A1, respectively.