accessibility menu, dialog, popup

UserName

CAS RN 459-57-4

FO

CAS RN 459-57-4

IUPAC Name: 4-fluorobenzaldehyde
Synonyms: benzaldehyde, 4-fluoro-
Molecular Weight (g/mol): 124.11
Molecular Formula: C7H5FO
InChi Key: UOQXIWFBQSVDPP-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)
  • (4)