missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzaldehyde, 98+%

Product Code. 10605022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10605022 250 mL Glass bottle 250mL
10533901 50 mL Glass bottle 50mL
10339093 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10605022 Supplier Thermo Scientific Acros Supplier No. 156152500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 459-57-4 | C7H5FO | 124.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-57-4
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00003378
InChI Key UOQXIWFBQSVDPP-UHFFFAOYSA-N
Synonym p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde
PubChem CID 68023
IUPAC Name 4-fluorobenzaldehyde
SMILES C1=CC(=CC=C1C=O)F

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -10.0°C
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 181.0°C (758.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CHO
Refractive Index 1.5195 to 1.5215
Quantity 250 mL
Beilstein 07, I, 132
Specific Gravity 1.15
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ether,acetone,benzene and alcohol
Formula Weight 124.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzaldehyde, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Fluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.