CAS RN 501-36-0
CAS RN 501-36-0
IUPAC Name:
5-[(1E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
Synonyms:
resveratrol trans-resveratrol 3,4',5-trihydroxystilbene 3,4',5-stilbenetriol resveratrol (substance) RESV resveratrol-3-sulfate (E)-resveratrol 3,4',5-trihydroxy-stilbene
Molecular Weight (g/mol):
228.25
Molecular Formula:
C14H12O3
InChi Key:
LUKBXSAWLPMMSZ-OWOJBTEDSA-N
SMILES:
OC1=CC=C(\C=C\C2=CC(O)=CC(O)=C2)C=C1
1
–
5
of
5
results
4
5
Resveratrol, MedChemExpress
MedChemExpress Resveratrol (trans-Resveratrol; SRT501), a natural polyphenolic phytoalexin that possesses anti-oxidant, anti-inflammatory, cardioprotective, and anti-cancer properties.