CAS RN 50767-79-8
CAS RN 50767-79-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(9Z,11E)-Prodlure, MedChemExpress
MedChemExpress (9Z,11E)-Prodlure ((9Z,11E)-Tetradecadien-1-yl acetate) is the main component of the sex pheromone of female Spodoptera littoralis.