accessibility menu, dialog, popup

UserName

CH3OOBrH2N

CAS RN 52727-57-8

IUPAC Name: methyl 2-amino-5-bromobenzoate
Synonyms: methyl 2-amino-5-bromobenzoate
Molecular Weight (g/mol): 230.06
Molecular Formula: C8H8BrNO2
InChi Key: QVNYNHCNNGKULA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CC=C1N
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results