missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-bromobenzoate, 98%

Product Code. 11452648
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452648 100 g 100g
11432648 5 g 5g
11442648 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452648 Supplier Thermo Scientific Alfa Aesar Supplier No. A16666.22
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52727-57-8 | C8H8BrNO2 | 230.06 g/mol

Methyl 2-amino-5-bromobenzoate acts as a potential PqsD inhibitor against pseudomonas aeruginosa infections. It is also used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. It is involved in plant growth-regulator activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-amino-5-bromobenzoate acts as a potential PqsD inhibitor against pseudomonas aeruginosa infections. It is also used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. It is involved in plant growth-regulator activity.

Solubility
Soluble in water.

Notes
Light and air sensitive. Incompatible with strong oxidizing agents, strong reducing agents, acid halides, acid anhydrides, strong acids and strong bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52727-57-8
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD00144761
InChI Key QVNYNHCNNGKULA-UHFFFAOYSA-N
Synonym methyl 5-bromoanthranilate, 2-amino-5-bromobenzoic acid methyl ester, 5-bromoanthranilic acid methyl ester, 2-amino-5-bromo-benzoic acid methyl ester, methyl 5-bromo-2-aminobenzoate, benzoic acid, 2-amino-5-bromo-, methyl ester, benzoic acid, 2-amino-5-bromo-, methyl ester, hydrochloride, pubchem3344, methyl-5-bromoanthranilate, acmc-1ats7
PubChem CID 736224
IUPAC Name methyl 2-amino-5-bromobenzoate
SMILES COC(=O)C1=CC(Br)=CC=C1N

Specifications

Melting Point 71°C to 73°C
Quantity 100 g
Beilstein 972720
Solubility Information Soluble in water.
Formula Weight 230.06
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-5-bromobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.