accessibility menu, dialog, popup

UserName

OH(R)O(S)OOHOOOH(R)OH(S)OH(S)OH

CAS RN 529-59-9

IUPAC Name: 5-hydroxy-3-(4-hydroxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4H-chromen-4-one
Synonyms: genistin genisteol 7-monoglucoside genistein 7-o-glucoside genistein glucoside genistine genistein 7-O-β-D-glucoside genistein-7-glucoside genistoside genistein, 7-O-β-D-glucoside 7-O-β-D-glucoside genistein
Molecular Weight (g/mol): 432.38
Molecular Formula: C21H20O10
InChi Key: ZCOLJUOHXJRHDI-CMWLGVBASA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC(O)=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol) 
  • (8)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (5)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (8)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (5)