missing translation for 'onlineSavingsMsg'
Learn More

Genistin, TRC

Product Code. 30004386
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004386 25 mg 25mg
17929074 10 mg 10mg
17999054 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004386 Supplier Toronto Research Chemicals Supplier No. TRCG35002025MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-59-9
Molecular Formula C21 H20 O10
Molecular Weight (g/mol) 432.38
Synonym 7-(β-D-Glucopyranosyloxy)-5-hydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Genistin (6CI,7CI,8CI), 7-(β-D-Glucopyranosyloxy)-5-hydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',5,7-Trihydroxyisoflavone 7-β-D-glucopyranoside, 5,4'-Di-hydroxyisoflavone-7-O-β-D-glucopyranoside, Genistein 7-O-glucoside, Genistein 7-O-β-D-glucopyranoside, Genistein 7-O-β-D-glucoside, Genistein 7-O-β-glucoside, Genistein, 7-β-D-glucopyranoside, Genisteol 7-monoglucoside, Genistine, Genistoside, NSC 5112
IUPAC Name 5-hydroxy-3-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
SMILES OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C(=COc3c2)c4ccc(O)cc4)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H20O10/c22-7-15-18(26)19(27)20(28)21(31-15)30-11-5-13(24)16-14(6-11)29-8-12(17(16)25)9-1-3-10(23)4-2-9/h1-6,8,15,18-24,26-28H,7H2/t15-,18-,19+,20-,21-/m1/s1
Formula Weight 432.1056
Percent Purity >95%
Chemical Name or Material Genistin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.