accessibility menu, dialog, popup

UserName

CAS RN 535-80-8

IUPAC Name: 3-chlorobenzoic acid
Synonyms: 3-chlorobenzoic acid 3-cholro-benzoic acid acido m-clorobenzoico C7H5ClO2 M-chlorobenzoic acid
Molecular Weight (g/mol): 156.57
Molecular Formula: C7H5ClO2
InChi Key: LULAYUGMBFYYEX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (4)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)