accessibility menu, dialog, popup

UserName

NNNN

CAS RN 54-95-5

IUPAC Name: 5H,6H,7H,8H,9H-[1,2,3,4]tetrazolo[1,5-a]azepine
Synonyms: cardiol pentamethylene-tetrazole metalex-P tetrazole, (1,5-pentanediyl)- corazole (analeptic) pentylenetetrazol preparation cerebro-nicin pentetrazolo pentrazole pentylenetetrazole
Molecular Weight (g/mol): 138.17
Molecular Formula: C6H10N4
InChi Key: CWRVKFFCRWGWCS-UHFFFAOYSA-N
SMILES: C1CCN2N=NN=C2CC1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results