accessibility menu, dialog, popup

UserName

H3CNCH3NHOOCH3NH2ClH2OHCl

CAS RN 54143-57-6

IUPAC Name: 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride
Synonyms: reglan pramiel gastrosil gastrotem gastromax mygdalon duraclamid peraprin metocobil metamide
Molecular Weight (g/mol): 354.27
Molecular Formula: C14H25Cl2N3O3
InChi Key: KJBLQGHJOCAOJP-UHFFFAOYSA-N
SMILES: O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results