missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxysalicylate, 98%, Thermo Scientific™

Product Code. 10046393 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10046393 50 g Glass bottle 50g
10285890 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10046393 Supplier Thermo Scientific Acros Supplier No. 174270500
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5446-02-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00008424
InChI Key ZICRWXFGZCVTBZ-UHFFFAOYSA-N
Synonym Methyle2-hydroxy-4-methoxybenzoate
IUPAC Name methyl 2-hydroxy-4-methoxybenzoate
SMILES COC(=O)C1=CC=C(OC)C=C1O

Specifications

Melting Point 49°C to 53°C
Color White to Yellow
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(OH)CO2CH3
Quantity 50 g
Beilstein 10, 381
Formula Weight 182.18
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl 4-methoxysalicylate
Show More Show Less

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.