accessibility menu, dialog, popup

UserName

CAS RN 55-38-9

IUPAC Name: O,O-dimethyl O-3-methyl-4-(methylsulfanyl)phenyl phosphorothioate
Synonyms: fenthion mpp (pesticide) queletox mercaptophos phenthion caswell no. 456F fenthion-methyl fenthion 4E fenthione MPP
Molecular Weight (g/mol): 278.32
Molecular Formula: C10H15O3PS2
InChi Key: PNVJTZOFSHSLTO-UHFFFAOYSA-N
SMILES: COP(=S)(OC)OC1=CC=C(SC)C(C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (1)
Concentration 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)