accessibility menu, dialog, popup

UserName

H3COOOOOCH3

CAS RN 56424-77-2

IUPAC Name: ethyl 2-(2-ethoxy-2-oxoethoxy)benzoate
Synonyms: ethyl 2-(2-ethoxy-2-oxoethoxy)benzoate
Molecular Weight (g/mol): 252.27
Molecular Formula: C13H16O5
InChi Key: NIUVYKIVCCMXLR-UHFFFAOYSA-N
SMILES: CCOC(=O)COC1=CC=CC=C1C(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results