accessibility menu, dialog, popup

UserName

HClNH2OFFFFF

CAS RN 57981-02-9

IUPAC Name: hydrogen O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine chloride
Synonyms: hydrogen O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine chloride
Molecular Weight (g/mol): 249.57
Molecular Formula: C7H5ClF5NO
InChi Key: HVMVKNXIMUCYJA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results