accessibility menu, dialog, popup

UserName

CAS RN 61281-38-7

H3COH3CCH3OCH3OCH3OCH3OH3COH3C(?)

CAS RN 61281-38-7

IUPAC Name: 3,4,5,14,15,16-hexamethoxy-9,10-dimethyltricyclo[10.4.0.0²,⁷]hexadeca-1(12),2,4,6,13,15-hexaene
Synonyms: 3,4,5,14,15,16-hexamethoxy-9,10-dimethyltricyclo[10.4.0.0²,⁷]hexadeca-1(12),2,4,6,13,15-hexaene
Molecular Weight (g/mol): 416.51
Molecular Formula: C24H32O6
InChi Key: JEJFTTRHGBKKEI-UHFFFAOYNA-N
SMILES: COC1=CC2=C(C(OC)=C1OC)C1=C(OC)C(OC)=C(OC)C=C1CC(C)C(C)C2
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results