CAS RN 61643-23-0
CAS RN 61643-23-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Leflunomide 3-Isomer, TRC
CAS: 61643-23-0 Molecular Formula: C12 H9 F3 N2 O2 Molecular Weight (g/mol): 270.21 Synonym: 5-Methyl-N-[3-(trifluoromethyl)phenyl]isoxazole-4-carboxamide,4-Isoxazolecarboxamide, 5-methyl-N-[3-(trifluoromethyl)phenyl]- (9CI),Leflunomide 3-Isomer,N-(3'-Trifluoromethylphenyl)-5-methylisoxazole-4-carboxamide IUPAC Name: 5-methyl-N-[3-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide SMILES: Cc1oncc1C(=O)Nc2cccc(c2)C(F)(F)F
2
5-Methyl-N-[3-(trifluoromethyl)phenyl]isoxazole-4-carboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.