accessibility menu, dialog, popup

UserName

CAS RN 6205-08-9

H3CONHNH2OHO

CAS RN 6205-08-9

IUPAC Name: 5-amino-2-acetamidopentanoic acid
Synonyms: 5-amino-2-acetamidopentanoic acid
Molecular Weight (g/mol): 174.2
Molecular Formula: C7H14N2O3
InChi Key: JRLGPAXAGHMNOL-UHFFFAOYNA-N
SMILES: CC(=O)NC(CCCN)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results