accessibility menu, dialog, popup

UserName

CAS RN 62613-82-5

NH2ONOHO

CAS RN 62613-82-5

IUPAC Name: 2-(4-hydroxy-2-oxopyrrolidin-1-yl)acetamide
Synonyms: oxiracetam
Molecular Weight (g/mol): 158.16
Molecular Formula: C6H10N2O3
InChi Key: IHLAQQPQKRMGSS-UHFFFAOYNA-N
SMILES: NC(=O)CN1CC(O)CC1=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results