CAS RN 64-85-7
IUPAC Name:
(1S,3aS,3bS,9aR,9bS,11aS)-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms:
11-deoxycorticosterone desoxycorticosteronum 21 hydroxy 4 pregnene 3,20 dione desoxicortona desoxicortonum D4-pregnene-21-ol-3,20-dione reichstein substance Q 11-dehydroxycorticosterone DOC Doc
Molecular Weight (g/mol):
330.47
Molecular Formula:
C21H30O3
InChi Key:
ZESRJSPZRDMNHY-YFWFAHHUSA-N
SMILES:
C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)CO
1
–
4
of
4
results
1
11-Deoxy Corticosterone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
11-Deoxy Corticosterone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
11-Deoxy Corticosterone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Deoxycorticosterone, MedChemExpress
MedChemExpress Deoxycorticosterone is a steroid hormone produced by the adrenal gland that possesses mineralocorticoid activity and acts as an aldosterone precursor.