accessibility menu, dialog, popup

UserName

CAS RN 65896-11-9

BrH2NF

CAS RN 65896-11-9

IUPAC Name: 2-bromo-6-fluoroaniline
Synonyms: 2-bromo-6-fluoroaniline
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: ALZFPYUPNVLVQM-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)