missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluoroaniline, 95%, Thermo Scientific™

Product Code. 10396684 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10396684 5 mL Glass bottle 5mL
10214824 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396684 Supplier Thermo Scientific Acros Supplier No. 398880050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 65896-11-9
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.02
MDL Number MFCD01310982
InChI Key ALZFPYUPNVLVQM-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-6-fluoro, 2-bromo-6-fluoro-phenylamine, 2-bromo-6-fluorobenzenamine, 2-bromo-6-fluoro-aniline, 2-bromo-6-fluoro aniline, 2-bromo-6-fluorophenylamine, pubchem2923, acmc-209nsu, 2-bromo-6-flouroaniline, ksc493o8h
PubChem CID 2782940
IUPAC Name 2-bromo-6-fluoroaniline
SMILES NC1=C(F)C=CC=C1Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.6740g/mL
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(F)NH2
Refractive Index 1.5810 to 1.5830 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.674
Formula Weight 190.01
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Bromo-6-fluoroaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-6-fluoroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.