CAS RN 68432-95-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-[(1RS)-1-Carboxyethyl]benzoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3-Carboxy-alpha-methylbenzeneacetic Acid(Ketoprofen Impurity), TRC
CAS: 68432-95-1 Molecular Formula: C10 H10 O4 Molecular Weight (g/mol): 194.18 Synonym: 3-[(1RS)-1-Carboxyethyl]benzoic acid,Ketoprofen Imp. C (EP),Benzeneacetic acid, 3-carboxy-α-methyl-,3-Carboxy-α-methylbenzeneacetic acid,3-(1-Carboxyethyl)benzoic acid,DF 2008Y IUPAC Name: 3-(1-carboxyethyl)benzoic acid SMILES: CC(C(=O)O)c1cccc(c1)C(=O)O