CAS RN 68524-30-1
CAS RN 68524-30-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Deoxy Modafinil, TRC
CAS: 68524-30-1 Molecular Formula: C15 H15 N O S Molecular Weight (g/mol): 257.35 Synonym: 2-[(Diphenylmethyl)sulfenyl]acetamide IUPAC Name: 2-benzhydrylsulfanylacetamide SMILES: NC(=O)CSC(c1ccccc1)c2ccccc2
2
2-[(Diphenylmethyl)sulfenyl]acetamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.