accessibility menu, dialog, popup

UserName

H3NFF

CAS RN 69385-30-4

IUPAC Name: (2,6-difluorophenyl)methanaminium
Synonyms: (2,6-difluorophenyl)methanaminium
Molecular Weight (g/mol): 144.14
Molecular Formula: C7H8F2N
InChi Key: PQCUDKMMPTXMAL-UHFFFAOYSA-O
SMILES: [NH3+]CC1=C(F)C=CC=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results