missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzylamine, 97+%, Thermo Scientific™

Product Code. 11443387
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11443387 5 g 5g
11433387 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11443387

Brand: Thermo Scientific Alfa Aesar A17169.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69385-30-4
Molecular Formula C7H8F2N
Molecular Weight (g/mol) 144.14
MDL Number MFCD00010144
InChI Key PQCUDKMMPTXMAL-UHFFFAOYSA-O
PubChem CID 123563
SMILES [NH3+]CC1=C(F)C=CC=C1F

Specifications

Density 1.197
Boiling Point 174°C to 177°C
Flash Point 65°C (149°F)
Refractive Index 1.493
Quantity 5 g
UN Number UN2735
Beilstein 2716771
Sensitivity Air Sensitive
Synonym 2,6-difluorobenzylamine, 2,6-difluorophenyl methanamine, 2,6-difluorobenzyl amine, benzenemethanamine, 2,6-difluoro, 1-2,6-difluorophenyl methanamine, 2,6-difluorophenyl methylamine, 2, 6-difluorobenzylamine, pubchem2235, 2,6-difluoro-benzylamine, acmc-1b2iy
IUPAC Name (2,6-difluorophenyl)methanaminium
Formula Weight 143.14
Percent Purity ≥97%
Chemical Name or Material 2,6-Difluorobenzylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.