missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzylamine, 97%, ACROS Organics™

Product Code. 10743203 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10743203 5 g Glass bottle 5g
10297550 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10743203

Brand: Thermo Scientific Acros 303590050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 69385-30-4
Molecular Formula C7H8F2N
Molecular Weight (g/mol) 144.14
MDL Number MFCD00010144
InChI Key PQCUDKMMPTXMAL-UHFFFAOYSA-O
PubChem CID 123563
IUPAC Name (2,6-difluorophenyl)methanaminium
SMILES [NH3+]CC1=C(F)C=CC=C1F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1900g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula F2C6H3CH2NH2
Refractive Index 1.4920 to 1.4940 (20°C, 589nm)
Quantity 5 g
Synonym 2,6-difluorobenzylamine, 2,6-difluorophenyl methanamine, 2,6-difluorobenzyl amine, benzenemethanamine, 2,6-difluoro, 1-2,6-difluorophenyl methanamine, 2,6-difluorophenyl methylamine, 2, 6-difluorobenzylamine, pubchem2235, 2,6-difluoro-benzylamine, acmc-1b2iy
Formula Weight 143.14
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 2, 6-Difluorobenzylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.