accessibility menu, dialog, popup

UserName

H3COONHNH2N

CAS RN 6994-25-8

IUPAC Name: ethyl 5-amino-1H-pyrazole-4-carboxylate
Synonyms: ethyl 3-amino-2H-pyrazole-4-carboxylate
Molecular Weight (g/mol): 155.16
Molecular Formula: C6H9N3O2
InChi Key: YPXGHKWOJXQLQU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)NN=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results