CAS RN 71-63-6
CAS RN 71-63-6
IUPAC Name:
4-[(1R,3aS,3bR,5aR,7S,9aS,9bS,11aR)-7-{[(2R,4S,5S,6R)-5-{[(2S,4S,5S,6R)-5-{[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-3a-hydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one
Synonyms:
digitoxin digitossina asthenthilo crystalline purodigin corax brand of digitoxin purodigin, crystalline cristapurat caswell no. 349 purodigin crystodigin
Molecular Weight (g/mol):
764.95
Molecular Formula:
C41H64O13
InChi Key:
WDJUZGPOPHTGOT-XUDUSOBPSA-N
SMILES:
C[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]1[C@@H](O)C[C@H](O[C@H]2[C@@H](O)C[C@H](O[C@H]3CC[C@@]4(C)[C@H](CC[C@@H]5[C@@H]4CC[C@]4(C)[C@H](CC[C@]54O)C4=CC(=O)OC4)C3)O[C@@H]2C)O[C@@H]1C
1
–
2
of
2
results
1
Digitoxin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
MedChemExpress Digitoxin
MedChemExpress Digitoxin is an effective Na+/K+-ATPase inhibitor, the EC50 value of Digitoxin is 0.78 μM.