missing translation for 'onlineSavingsMsg'
Learn More

Digitoxin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30315645
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30315645

Brand: Dr. Ehrenstorfer DREA12633800AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Digitoxin
CAS 71-63-6
Analyte or Component Names Digitoxin
Shipping Condition Room Temperature
Molecular Formula C41 H64 O13
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Digitoxin, 3β-[(O-2,6-dideoxy-β-d-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-d-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-d-ribo-hexopyranosyl)oxy]-14-hydroxy-5β,14β-card-20(22)-enolide, Card-20(22)-enolide, 3-[(O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-14-hydroxy-, (3β,5β)-, Digitoxin (8CI), Digitoxoside (7CI), (3β,5β)-3-[(O-2,6-Dideoxy-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-14-hydroxycard-20(22)-enolide, Cardidigin, Cardigin, Carditoxin, Coramedan, Cristapurat, Crystalline digitalin, Crystodigin, Digicor, Digilong, Digimed, Digimerck, Digipural, Digisidin, Digitaline Nativelle, Digitophyllin, Digitoxigenin tridigitoxoside, Digitrin, Ditaven, Glucodigin, Lanatoxin, Myodigin, NSC 7529, Purodigin, Purpurid, Tradigal, Unidigin
InChI Formula InChI=1S/C41H64O13/c1-20-36(46)29(42)16-34(49-20)53-38-22(3)51-35(18-31(38)44)54-37-21(2)50-33(17-30(37)43)52-25-8-11-39(4)24(15-25)6-7-28-27(39)9-12-40(5)26(10-13-41(28,40)47)23-14-32(45)48-19-23/h14,20-22,24-31,33-38,42-44,46-47H,6-13,15-19H2,1-5H3/t20-,21-,22-,24-,25+,26-,27+,28-,29+,30+,31+,33+,34+,35+,36-,37-,38-,39+,40-,41+/m1/s1
SMILES C[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]2[C@@H](O)C[C@H](O[C@H]3[C@@H](O)C[C@H](O[C@H]4CC[C@@]5(C)[C@H](CC[C@@H]6[C@@H]5CC[C@]7(C)[C@H](CC[C@]67O)C8=CC(=O)OC8)C4)O[C@@H]3C)O[C@@H]2C
IUPAC Name 3-[(3S,5R,8R,9S,10S,13R,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-14-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2H-furan-5-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 764.94
Quantity 1 mL
Formula Weight 764.4347 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.