accessibility menu, dialog, popup

UserName

CAS RN 72505-21-6

H2NSFFF

CAS RN 72505-21-6

IUPAC Name: 4-(trifluoromethyl)benzene-1-carbothioamide
Synonyms: 4-(trifluoromethyl)benzenecarbothioamide
Molecular Weight (g/mol): 205.2
Molecular Formula: C8H6F3NS
InChi Key: IPRFNMJROWWFBH-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (4)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)