accessibility menu, dialog, popup

UserName

SNNOOOHONa

CAS RN 75277-39-3

IUPAC Name: sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Synonyms: hydroxyethylpiperazine ethane sulfonic acid
Molecular Weight (g/mol): 260.28
Molecular Formula: C8H17N2NaO4S
InChi Key: RDZTWEVXRGYCFV-UHFFFAOYSA-M
SMILES: [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
Quantity 
  • (4)
  • (1)
  • (6)
  • (4)
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
  • (5)
  • (4)
  • (2)
Physical Form 
  • (6)
  • (8)
  • (1)
Grade 
  • (5)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (8)
  • (6)
  • (2)
  • (3)
  • (4)
  • (1)
  • (6)
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (5)
  • (4)
  • (2)
  • (6)
  • (8)
  • (1)
  • (5)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (4)
  • (6)
  • (2)
  • (2)