missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES sodium salt, 99%

Product Code. 10542714 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10542714 500 g 500g
10695852 25 g 25g
10411451 100 g 100g
10532554 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10542714 Supplier Thermo Scientific Chemicals Supplier No. 215005000
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75277-39-3 | C8H17N2NaO4S | 260.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Absorbance (0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm, 0.02 at 280nm, 0.04 at 260nm
Absorbance Conditions (0.1M aq. soln.)
Color White
pH 9.5 to 10.5 (1% aq. soln.)
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Formula Weight 260.27
Percent Purity ≥98.5% (on anhydrous substance)
Chemical Name or Material HEPES sodium salt, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.