missing translation for 'onlineSavingsMsg'
Learn More

HEPES Sodium Salt (White Powder), Fisher BioReagents

Product Code. 10193293
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10193293 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10193293 Supplier Fisher BioReagents Supplier No. BP4101
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
HEPES Sodium Salt is used as a buffer for in vitro cell culture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Absorbance 0.04 max. (0.1M in water, 1cm) at 260nm, 0.02 max. (0.1M in water, 1cm) at 280nm
Color White
Physical Form Powder or Solid
Quantity 1 kg
Assay Percent Range ≥99 %
Identification Pass Test
Loss on Drying 3.0% max.
Packaging Poly Bottle
Formula Weight 260.29
Percent Purity ≥99%
Chemical Name or Material HEPES Sodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.