accessibility menu, dialog, popup

UserName

CAS RN 75507-68-5

H3COOHNNHFNH2NOHOOHO(Z)

CAS RN 75507-68-5

IUPAC Name: (2Z)-but-2-enedioic acid; ethyl N-(2-amino-6-{[(4-fluorophenyl)methyl]amino}pyridin-3-yl)carbamate
Synonyms: flupirtine
Molecular Weight (g/mol): 420.4
Molecular Formula: C19H21FN4O6
InChi Key: DPYIXBFZUMCMJM-BTJKTKAUSA-N
SMILES: OC(=O)\C=C/C(O)=O.CCOC(=O)NC1=CC=C(NCC2=CC=C(F)C=C2)N=C1N
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results