missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flupirtine Maleate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Flupirtine Maleate
Specifications
Specifications
| Chemical Name or Material | Flupirtine Maleate |
| CAS | 75507-68-5 |
| Analyte or Component Names | Flupirtine Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H17 F N4 O2 . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Flupirtine Maleate, Carbamic acid, N-[2-amino-6-[[(4-fluorophenyl)methyl]amino]-3-pyridinyl]-, ethyl ester, (2Z)-2-butenedioate (1:1), Carbamic acid, [2-amino-6-[[(4-fluorophenyl)methyl]amino]-3-pyridinyl]-, ethyl ester, (2Z)-2-butenedioate (1:1) (9CI), Carbamic acid, [2-amino-6-[[(4-fluorophenyl)methyl]amino]-3-pyridinyl]-, ethyl ester, (Z)-2-butenedioate (1:1), Efiret, W 2964M |
| InChI Formula | InChI=1S/C15H17FN4O2.C4H4O4/c1-2-22-15(21)19-12-7-8-13(20-14(12)17)18-9-10-3-5-11(16)6-4-10;5-3(6)1-2-4(7)8/h3-8H,2,9H2,1H3,(H,19,21)(H3,17,18,20);1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CCOC(=O)Nc1ccc(NCc2ccc(F)cc2)nc1N.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;ethyl N-[2-amino-6-[(4-fluorophenyl)methylamino]pyridin-3-yl]carbamate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction